Learn More
4-Chloro-N-methylaniline, 95%, Thermo Scientific Chemicals
£16.68 - £45.12
Chemical Identifiers
CAS | 932-96-7 |
---|---|
Molecular Formula | C7H8ClN |
Molecular Weight (g/mol) | 141.598 |
MDL Number | MFCD00000614 |
InChI Key | XCEYKKJMLOFDSS-UHFFFAOYSA-N |
Synonym | p-chloro-n-methylaniline, benzenamine, 4-chloro-n-methyl, n-methyl-4-chloroaniline, n-p-chlorobenzyl methylamine, n-methyl-p-chloroaniline, aniline, p-chloro-n-methyl, unii-2ixy9ja2p8, 4-chloro-n-methylbenzenamine, ccris 2889, n-4-chlorophenyl-n-methylamine |
PubChem CID | 70272 |
IUPAC Name | 4-chloro-N-methylaniline |
SMILES | CNC1=CC=C(C=C1)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11496836
|
Thermo Scientific Alfa Aesar
B23962.03 |
1 g |
£16.68
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11406846
|
Thermo Scientific Alfa Aesar
B23962.06 |
5 g |
£45.12
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
932-96-7 | |
141.598 | |
XCEYKKJMLOFDSS-UHFFFAOYSA-N | |
70272 | |
CNC1=CC=C(C=C1)Cl |
C7H8ClN | |
MFCD00000614 | |
p-chloro-n-methylaniline, benzenamine, 4-chloro-n-methyl, n-methyl-4-chloroaniline, n-p-chlorobenzyl methylamine, n-methyl-p-chloroaniline, aniline, p-chloro-n-methyl, unii-2ixy9ja2p8, 4-chloro-n-methylbenzenamine, ccris 2889, n-4-chlorophenyl-n-methylamine | |
4-chloro-N-methylaniline |
Specifications
932-96-7 | |
>100°C (212°F) | |
MFCD00000614 | |
2205846 | |
XCEYKKJMLOFDSS-UHFFFAOYSA-N | |
4-chloro-N-methylaniline | |
70272 | |
95% |
238°C to 240°C | |
C7H8ClN | |
UN2810 | |
p-chloro-n-methylaniline, benzenamine, 4-chloro-n-methyl, n-methyl-4-chloroaniline, n-p-chlorobenzyl methylamine, n-methyl-p-chloroaniline, aniline, p-chloro-n-methyl, unii-2ixy9ja2p8, 4-chloro-n-methylbenzenamine, ccris 2889, n-4-chlorophenyl-n-methylamine | |
CNC1=CC=C(C=C1)Cl | |
141.598 | |
141.6 | |
4-Chloro-N-methylaniline |
Safety and Handling
GHS H Statement
H331-H373-H302-H312-H315-H319
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
P260-P261-P280-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
missing translation for 'rtecsNumber' : CX9857900
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.