missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-N-methylaniline, 95%, Thermo Scientific Chemicals

Product Code. 11406846
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 932-96-7
Molecular Formula C7H8ClN
Molecular Weight (g/mol) 141.598
MDL Number MFCD00000614
InChI Key XCEYKKJMLOFDSS-UHFFFAOYSA-N
Synonym p-chloro-n-methylaniline, benzenamine, 4-chloro-n-methyl, n-methyl-4-chloroaniline, n-p-chlorobenzyl methylamine, n-methyl-p-chloroaniline, aniline, p-chloro-n-methyl, unii-2ixy9ja2p8, 4-chloro-n-methylbenzenamine, ccris 2889, n-4-chlorophenyl-n-methylamine
PubChem CID 70272
IUPAC Name 4-chloro-N-methylaniline
SMILES CNC1=CC=C(C=C1)Cl

Specifications

Density 1.169
Boiling Point 238°C to 240°C
Flash Point >100°C (212°F)
Refractive Index 1.584
Quantity 5 g
UN Number UN2810
Beilstein 2205846
Formula Weight 141.6
Percent Purity 95%
Chemical Name or Material 4-Chloro-N-methylaniline

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'