missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethoxybenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals

£61.20 - £900.00

Chemical Identifiers

CAS 23095-31-0
Molecular Formula C8H9ClO4S
Molecular Weight (g/mol) 236.68
MDL Number MFCD00051769
InChI Key RSJSYCZYQNJQPY-UHFFFAOYSA-N
Synonym 3,4-dimethoxybenzene-1-sulfonyl chloride, 3,4-dimethoxybenzenesulfonylchloride, 3,4-dimethoxyphenylsulfonyl chloride, 3,4-dimethoxy benzenesulfonyl chloride, benzenesulfonyl chloride, 3,4-dimethoxy, 3,4-dimethoxyphenyl chlorosulfone, pubchem5127, acmc-1co88, asischem d48920, zerenex zx009571
PubChem CID 2734183
IUPAC Name 3,4-dimethoxybenzenesulfonyl chloride
SMILES COC1=C(C=C(C=C1)S(=O)(=O)Cl)OC
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10363962
View Documents
Thermo Scientific Acros
354390010
1 g Glass bottle
£61.20
1g
Please to purchase this item. Need a web account? Register with us today!
10652223
View Documents
Thermo Scientific Acros
354390050
5 g Glass bottle
£205.02
5g
Please to purchase this item. Need a web account? Register with us today!
10095863
View Documents
Thermo Scientific Acros
354390250
25 g Glass bottle
£900.00
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

23095-31-0
236.68
RSJSYCZYQNJQPY-UHFFFAOYSA-N
2734183
COC1=C(C=C(C=C1)S(=O)(=O)Cl)OC
C8H9ClO4S
MFCD00051769
3,4-dimethoxybenzene-1-sulfonyl chloride, 3,4-dimethoxybenzenesulfonylchloride, 3,4-dimethoxyphenylsulfonyl chloride, 3,4-dimethoxy benzenesulfonyl chloride, benzenesulfonyl chloride, 3,4-dimethoxy, 3,4-dimethoxyphenyl chlorosulfone, pubchem5127, acmc-1co88, asischem d48920, zerenex zx009571
3,4-dimethoxybenzenesulfonyl chloride

Specifications

23095-31-0
100.0
White to Beige
96% min. (HPLC)
(CH3O)2C6H3SO2Cl
3,4-dimethoxybenzene-1-sulfonyl chloride, 3,4-dimethoxybenzenesulfonylchloride, 3,4-dimethoxyphenylsulfonyl chloride, 3,4-dimethoxy benzenesulfonyl chloride, benzenesulfonyl chloride, 3,4-dimethoxy, 3,4-dimethoxyphenyl chlorosulfone, pubchem5127, acmc-1co88, asischem d48920, zerenex zx009571
COC1=C(C=C(C=C1)S(=O)(=O)Cl)OC
236.68
236.68
Crystalline Powder
96.0
65.0°C to 70.0°C
Authentic
C8H9ClO4S
MFCD00051769
RSJSYCZYQNJQPY-UHFFFAOYSA-N
3,4-dimethoxybenzenesulfonyl chloride
2734183
97%
3, 4-Dimethoxybenzenesulfonyl chloride, 97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

Product Identifier
  • 3,4-Dimethoxybenzenesulfonyl chloride

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only