missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethoxybenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals

Product Code. 10095863
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23095-31-0
Molecular Formula C8H9ClO4S
Molecular Weight (g/mol) 236.68
MDL Number MFCD00051769
InChI Key RSJSYCZYQNJQPY-UHFFFAOYSA-N
Synonym 3,4-dimethoxybenzene-1-sulfonyl chloride, 3,4-dimethoxybenzenesulfonylchloride, 3,4-dimethoxyphenylsulfonyl chloride, 3,4-dimethoxy benzenesulfonyl chloride, benzenesulfonyl chloride, 3,4-dimethoxy, 3,4-dimethoxyphenyl chlorosulfone, pubchem5127, acmc-1co88, asischem d48920, zerenex zx009571
PubChem CID 2734183
IUPAC Name 3,4-dimethoxybenzenesulfonyl chloride
SMILES COC1=C(C=C(C=C1)S(=O)(=O)Cl)OC

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 65.0°C to 70.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula (CH3O)2C6H3SO2Cl
Quantity 25 g
Formula Weight 236.68
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 3, 4-Dimethoxybenzenesulfonyl chloride, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 3,4-Dimethoxybenzenesulfonyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'