Learn More
Lenz Laborglasintrumente™ 2-Neck Adapters
Two parallel necks with two NS sockets and one NS cone
£42.45 - £44.45
Product Code | Brand | Cone Size | Standard Taper Joint No. | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Cone Size | Standard Taper Joint No. | Price | Quantity & Availability | ||||
10654392
|
Lenz Laborglasintrumente™
05010033 |
NS 24/29 | NS 24/29 |
£44.45
Each |
Estimated Shipment: 06-01-2025
Log in to see stock. |
||||
10418792
|
Lenz Laborglasintrumente™
05010014 |
NS 29/32 | NS 14/23 |
£42.45
Each |
Estimated Shipment: 06-01-2025
Log in to see stock. |
||||
Description
- Acc. to DIN 12594
- Made of DURAN™ tubing
Specifications
Duran Tubing | |
1 |