Learn More
Lenz Laborglasintrumente™ 2-Neck Adapters
Two parallel necks with two NS sockets and one NS cone
Brand:  Lenz Laborglasintrumente™ 05010033
 View more versions of this product
Product Code. 10654392
  • £44.45 / Each
Estimated Shipment: 07-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Acc. to DIN 12594
  • Made of DURAN™ tubing
Specifications
Specifications
NS 24/29
Duran Tubing
1
NS 24/29