missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Fluorobenzyl)-1,4-diazepane, 97%, Thermo Scientific™

£13.80 - £64.40

Chemical Identifiers

CAS 76141-89-4
Molecular Formula C12H17FN2
Molecular Weight (g/mol) 208.28
MDL Number MFCD00816588
InChI Key KZCHYFUMQQWGJK-UHFFFAOYSA-N
Synonym 1-4-fluorobenzyl-1,4-diazepane, 1-4-fluorobenzyl homopiperazine, 1-4-fluorophenyl methyl-1,4-diazepane, 1-4-fluoro-benzyl-1,4 diazepane, 1-4-fluorophenyl methyl-1,4-diazaperhydroepine, acmc-1bluy, 1-4-fluorobenzyl-1,4 diazepane
PubChem CID 2774433
IUPAC Name 1-[(4-fluorophenyl)methyl]-1,4-diazepane
SMILES C1CNCCN(C1)CC2=CC=C(C=C2)F
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10582452
SDS View Documents
Thermo Scientific Maybridge
AW00512DA
1 g
£13.80
1g
Please to purchase this item. Need a web account? Register with us today!
10764657
SDS View Documents
Thermo Scientific Maybridge
AW00512DE
5 g
£38.00
5g
Please to purchase this item. Need a web account? Register with us today!
10525675
SDS View Documents
Thermo Scientific Maybridge
AW00512EA
10 g
£64.40
10g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

76141-89-4
208.28
KZCHYFUMQQWGJK-UHFFFAOYSA-N
2774433
C1CNCCN(C1)CC2=CC=C(C=C2)F
C12H17FN2
MFCD00816588
1-4-fluorobenzyl-1,4-diazepane, 1-4-fluorobenzyl homopiperazine, 1-4-fluorophenyl methyl-1,4-diazepane, 1-4-fluoro-benzyl-1,4 diazepane, 1-4-fluorophenyl methyl-1,4-diazaperhydroepine, acmc-1bluy, 1-4-fluorobenzyl-1,4 diazepane
1-[(4-fluorophenyl)methyl]-1,4-diazepane

Specifications

76141-89-4
100.0
Amber glass bottle
MFCD00816588
KZCHYFUMQQWGJK-UHFFFAOYSA-N
1-[(4-fluorophenyl)methyl]-1,4-diazepane
2774433
95%
94.5
95%
C12H17FN2
1-4-fluorobenzyl-1,4-diazepane, 1-4-fluorobenzyl homopiperazine, 1-4-fluorophenyl methyl-1,4-diazepane, 1-4-fluoro-benzyl-1,4 diazepane, 1-4-fluorophenyl methyl-1,4-diazaperhydroepine, acmc-1bluy, 1-4-fluorobenzyl-1,4 diazepane
C1CNCCN(C1)CC2=CC=C(C=C2)F
208.28
208.28
1-(4-Fluorobenzyl)-1, 4-diazepane
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.