missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Fluorobenzyl)-1,4-diazepane, 97%, Thermo Scientific™

Product Code. 10525675
Click to view available options
Quantity:
1 g
5 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 76141-89-4
Molecular Formula C12H17FN2
Molecular Weight (g/mol) 208.28
MDL Number MFCD00816588
InChI Key KZCHYFUMQQWGJK-UHFFFAOYSA-N
Synonym 1-4-fluorobenzyl-1,4-diazepane, 1-4-fluorobenzyl homopiperazine, 1-4-fluorophenyl methyl-1,4-diazepane, 1-4-fluoro-benzyl-1,4 diazepane, 1-4-fluorophenyl methyl-1,4-diazaperhydroepine, acmc-1bluy, 1-4-fluorobenzyl-1,4 diazepane
PubChem CID 2774433
IUPAC Name 1-[(4-fluorophenyl)methyl]-1,4-diazepane
SMILES C1CNCCN(C1)CC2=CC=C(C=C2)F

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Quantity 10 g
Formula Weight 208.28
Percent Purity 95%
Chemical Name or Material 1-(4-Fluorobenzyl)-1, 4-diazepane
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.