missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dihydroxyanthraquinone, 96%, Thermo Scientific Chemicals

£20.55 - £97.89

Chemical Identifiers

CAS 81-64-1
Molecular Formula C14H8O4
Molecular Weight (g/mol) 240.21
MDL Number MFCD00001209
InChI Key GUEIZVNYDFNHJU-UHFFFAOYSA-N
Synonym quinizarin, 1,4-dihydroxyanthraquinone, quinizarine, chinizarin, solvent orange 86, 1,4-dihydroxyanthrachinon, smoke orange r, 1,4-dihydroxy-9,10-anthraquinone, macrolex orange gg, 9,10-anthracenedione, 1,4-dihydroxy
PubChem CID 6688
ChEBI CHEBI:37487
IUPAC Name 1,4-dihydroxyanthracene-9,10-dione
SMILES C1=CC=C2C(=C1)C(=O)C3=C(C=CC(=C3C2=O)O)O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10607682
SDS View Documents
Thermo Scientific Acros
132180050
5 g Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10081430
SDS View Documents
Thermo Scientific Acros
132181000
100 g Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10697672
SDS View Documents
Thermo Scientific Acros
132180010
1 kg Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

81-64-1
240.21
GUEIZVNYDFNHJU-UHFFFAOYSA-N
6688
1,4-dihydroxyanthracene-9,10-dione
C14H8O4
MFCD00001209
quinizarin, 1,4-dihydroxyanthraquinone, quinizarine, chinizarin, solvent orange 86, 1,4-dihydroxyanthrachinon, smoke orange r, 1,4-dihydroxy-9,10-anthraquinone, macrolex orange gg, 9,10-anthracenedione, 1,4-dihydroxy
CHEBI:37487
C1=CC=C2C(=C1)C(=O)C3=C(C=CC(=C3C2=O)O)O

Specifications

81-64-1
100.0
Brown-Red to Orange
222°C
96%
MFCD00001209
15, 8180
Solubility in water: <1g/L (20°C). Other solubilities: soluble in hot water,soluble in alcohol,ether,benzene,sulfuric,acid,potassium chloride,chloroform,acetone,,aqueous alkali and other organic solvents
C1=CC=C2C(=C1)C(=O)C3=C(C=CC(=C3C2=O)O)O
240.21
CHEBI:37487
96%
1, 4-Dihydroxyanthraquinone, 96%
95.0
191.0°C to 193.0°C
450.0°C
Authentic
C14H8O4
08, 450
quinizarin, 1,4-dihydroxyanthraquinone, quinizarine, chinizarin, solvent orange 86, 1,4-dihydroxyanthrachinon, smoke orange r, 1,4-dihydroxy-9,10-anthraquinone, macrolex orange gg, 9,10-anthracenedione, 1,4-dihydroxy
GUEIZVNYDFNHJU-UHFFFAOYSA-N
1,4-dihydroxyanthracene-9,10-dione
6688
240.21
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Very toxic to aquatic life with long lasting effects.

GHS P Statement
Avoid release to the environment.
Collect spillage.
Dispose of contents/container to an approved waste disposal plant.

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 201-368-7

missing translation for 'rtecsNumber' : CB6600000

missing translation for 'tsca' : TSCA

Product Identifier
  • 1,4-Dihydroxyanthraquinone

Signal Word
  • Warning

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1

Hazard Statement
  • H410-Very toxic to aquatic life with long lasting effects.
  • H400-Very toxic to aquatic life.

Precautionary Statement
  • P273-Avoid release to the environment.
  • P391-Collect spillage.
  • P501-Dispose of contents/container to .

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.