missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dihydroxyanthraquinone, 96%, Thermo Scientific Chemicals

Product Code. 10081430
Click to view available options
Quantity:
5 g
100 g
1 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81-64-1
Molecular Formula C14H8O4
Molecular Weight (g/mol) 240.21
MDL Number MFCD00001209
InChI Key GUEIZVNYDFNHJU-UHFFFAOYSA-N
Synonym quinizarin, 1,4-dihydroxyanthraquinone, quinizarine, chinizarin, solvent orange 86, 1,4-dihydroxyanthrachinon, smoke orange r, 1,4-dihydroxy-9,10-anthraquinone, macrolex orange gg, 9,10-anthracenedione, 1,4-dihydroxy
PubChem CID 6688
ChEBI CHEBI:37487
IUPAC Name 1,4-dihydroxyanthracene-9,10-dione
SMILES C1=CC=C2C(=C1)C(=O)C3=C(C=CC(=C3C2=O)O)O

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 191.0°C to 193.0°C
Color Brown-Red to Orange
Boiling Point 450.0°C
Flash Point 222°C
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Quantity 100 g
Beilstein 08, 450
Merck Index 15, 8180
Solubility Information Solubility in water: <1g/L (20°C). Other solubilities: soluble in hot water,soluble in alcohol,ether,benzene,sulfuric,acid,potassium chloride,chloroform,acetone,,aqueous alkali and other organic solvents
Formula Weight 240.21
Percent Purity 96%
Physical Form Crystalline Powder
Chemical Name or Material 1, 4-Dihydroxyanthraquinone, 96%
Show More Show Less
compliance-icons
Product Identifier
  • 1,4-Dihydroxyanthraquinone
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H410-Very toxic to aquatic life with long lasting effects.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P391-Collect spillage.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.