missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dibromooctafluorobutane, 98%, Thermo Scientific™

£32.56 - £93.03

Chemical Identifiers

CAS 335-48-8
Molecular Formula C4Br2F8
Molecular Weight (g/mol) 359.839
MDL Number MFCD00153112
InChI Key RWWUGYJWSVESJC-UHFFFAOYSA-N
Synonym 1,4-dibromooctafluorobutane, 1,4-dibromoperfluorobutane, butane, 1,4-dibromo-1,1,2,2,3,3,4,4-octafluoro, acmc-209i1o, 1,4-dibromo-1,1,2,2,3,3,4,4-octafluoro-butane, butane,1,4-dibromo-1,1,2,2,3,3,4,4-octafluoro, 1,4-bis bromanyl-1,1,2,2,3,3,4,4-octakis fluoranyl butane
PubChem CID 2736783
IUPAC Name 1,4-dibromo-1,1,2,2,3,3,4,4-octafluorobutane
SMILES C(C(C(F)(F)Br)(F)F)(C(F)(F)Br)(F)F
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11379365
SDS View Documents
Thermo Scientific Alfa Aesar
L16736.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11389365
SDS View Documents
Thermo Scientific Alfa Aesar
L16736.14
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

335-48-8
359.839
RWWUGYJWSVESJC-UHFFFAOYSA-N
2736783
C(C(C(F)(F)Br)(F)F)(C(F)(F)Br)(F)F
C4Br2F8
MFCD00153112
1,4-dibromooctafluorobutane, 1,4-dibromoperfluorobutane, butane, 1,4-dibromo-1,1,2,2,3,3,4,4-octafluoro, acmc-209i1o, 1,4-dibromo-1,1,2,2,3,3,4,4-octafluoro-butane, butane,1,4-dibromo-1,1,2,2,3,3,4,4-octafluoro, 1,4-bis bromanyl-1,1,2,2,3,3,4,4-octakis fluoranyl butane
1,4-dibromo-1,1,2,2,3,3,4,4-octafluorobutane

Specifications

335-48-8
96°C to 98°C
Br(CF2)4Br
MFCD00153112
1,4-dibromooctafluorobutane, 1,4-dibromoperfluorobutane, butane, 1,4-dibromo-1,1,2,2,3,3,4,4-octafluoro, acmc-209i1o, 1,4-dibromo-1,1,2,2,3,3,4,4-octafluoro-butane, butane,1,4-dibromo-1,1,2,2,3,3,4,4-octafluoro, 1,4-bis bromanyl-1,1,2,2,3,3,4,4-octakis fluoranyl butane
C(C(C(F)(F)Br)(F)F)(C(F)(F)Br)(F)F
359.839
359.84
1,4-Dibromooctafluorobutane
2.098
C4Br2F8
1.352
1777536
RWWUGYJWSVESJC-UHFFFAOYSA-N
1,4-dibromo-1,1,2,2,3,3,4,4-octafluorobutane
2736783
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.