missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dibromooctafluorobutane, 98%, Thermo Scientific™

Product Code. 11379365
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 335-48-8
Molecular Formula C4Br2F8
Molecular Weight (g/mol) 359.839
MDL Number MFCD00153112
InChI Key RWWUGYJWSVESJC-UHFFFAOYSA-N
Synonym 1,4-dibromooctafluorobutane, 1,4-dibromoperfluorobutane, butane, 1,4-dibromo-1,1,2,2,3,3,4,4-octafluoro, acmc-209i1o, 1,4-dibromo-1,1,2,2,3,3,4,4-octafluoro-butane, butane,1,4-dibromo-1,1,2,2,3,3,4,4-octafluoro, 1,4-bis bromanyl-1,1,2,2,3,3,4,4-octakis fluoranyl butane
PubChem CID 2736783
IUPAC Name 1,4-dibromo-1,1,2,2,3,3,4,4-octafluorobutane
SMILES C(C(C(F)(F)Br)(F)F)(C(F)(F)Br)(F)F

Specifications

Density 2.098
Boiling Point 96°C to 98°C
Linear Formula Br(CF2)4Br
Refractive Index 1.352
Quantity 5 g
Beilstein 1777536
Formula Weight 359.84
Percent Purity 98%
Chemical Name or Material 1,4-Dibromooctafluorobutane
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.