missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dibromo-3-methylbutane, 98%, Thermo Scientific Chemicals

£25.70 - £295.00

Chemical Identifiers

CAS 24443-15-0
Molecular Formula C5H10Br2
Molecular Weight (g/mol) 229.94
MDL Number MFCD00060773
InChI Key XMJFMQSXQQUJTQ-UHFFFAOYSA-N
Synonym butane, 1,3-dibromo-3-methyl, acmc-20ap4g, 2,4-dibromo-2-methylbutane
PubChem CID 141113
IUPAC Name 1,3-dibromo-3-methylbutane
SMILES CC(C)(Br)CCBr
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11444447
View Documents
Thermo Scientific Alfa Aesar
B20468.06
5 g
£25.70
5g
Please to purchase this item. Need a web account? Register with us today!
11454447
View Documents
Thermo Scientific Alfa Aesar
B20468.14
25 g
£83.60
25g
Please to purchase this item. Need a web account? Register with us today!
11464447
View Documents
Thermo Scientific Alfa Aesar
B20468.22
100 g
£295.00
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

24443-15-0
229.94
XMJFMQSXQQUJTQ-UHFFFAOYSA-N
141113
CC(C)(Br)CCBr
C5H10Br2
MFCD00060773
butane, 1,3-dibromo-3-methyl, acmc-20ap4g, 2,4-dibromo-2-methylbutane
1,3-dibromo-3-methylbutane

Specifications

24443-15-0
>110°C (230°F)
MFCD00060773
butane, 1,3-dibromo-3-methyl, acmc-20ap4g, 2,4-dibromo-2-methylbutane
CC(C)(Br)CCBr
229.94
229.95
1,3-Dibromo-3-methylbutane
1.69
C5H10Br2
Light sensitive
XMJFMQSXQQUJTQ-UHFFFAOYSA-N
1,3-dibromo-3-methylbutane
141113
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only