missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dibromo-3-methylbutane, 98%, Thermo Scientific Chemicals

Product Code. 11454447
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24443-15-0
Molecular Formula C5H10Br2
Molecular Weight (g/mol) 229.94
MDL Number MFCD00060773
InChI Key XMJFMQSXQQUJTQ-UHFFFAOYSA-N
Synonym butane, 1,3-dibromo-3-methyl, acmc-20ap4g, 2,4-dibromo-2-methylbutane
PubChem CID 141113
IUPAC Name 1,3-dibromo-3-methylbutane
SMILES CC(C)(Br)CCBr

Specifications

Density 1.69
Flash Point >110°C (230°F)
Quantity 25 g
Sensitivity Light sensitive
Formula Weight 229.95
Percent Purity 98%
Chemical Name or Material 1,3-Dibromo-3-methylbutane

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'