missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Bis(chloromethyl)tetramethyldisiloxane, 97%, Thermo Scientific™
£69.82
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11301587
|
Thermo Scientific Alfa Aesar
L04436.18 |
50 g | N/A | ||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications1,3-Bis(chloromethyl)tetramethyldisiloxane is a useful bifunctional silylating agent.
Solubility
Not miscible or difficult to mix with water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Chemical Identifiers
2362-10-9 | |
MFCD00000877 |
Specifications
92°C to 93°C (20mmHg) | |
C6H16Cl2OSi2 | |
506631 | |
1,3-bis chloromethyl-1,1,3,3-tetramethyldisiloxane, 1,3-bis chloromethyl tetramethyldisiloxane, bis chloromethyl tetramethyldisiloxane, disiloxane, 1,3-bis chloromethyl-1,1,3,3-tetramethyl, tetramethylbis chloromethyl disiloxane, tetramethyl-1,3-bis chloromethyl disiloxane, chloromethyl chloromethyl dimethylsilyl oxy dimethylsilane, disiloxane,3-bis chloromethyl-1,1,3,3-tetramethyl, pubchem13666, acmc-2097an | |
NBGGEWGFZUDQKZ-UHFFFAOYSA-N | |
chloromethyl-[chloromethyl(dimethyl)silyl]oxy-dimethylsilane | |
75385 | |
97% | |
1.05 |
2362-10-9 | |
MFCD00000877 | |
73°C (163°F) | |
Not miscible or difficult to mix with water. | |
C[Si](C)(CCl)O[Si](C)(C)CCl | |
231.26 | |
231.27 | |
1.44 | |
1,3-Bis(chloromethyl)tetramethyldisiloxane |
Safety and Handling
GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
P210-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c
H227-H315-H319
missing translation for 'einecsNumber' : 219-109-1
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only