missing translation for 'onlineSavingsMsg'
Learn More

1,3-Bis(chloromethyl)tetramethyldisiloxane, 97%, Thermo Scientific™

Product Code. 11301587
Click to view available options
Quantity:
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
1,3-Bis(chloromethyl)tetramethyldisiloxane is a useful bifunctional silylating agent.

Solubility
Not miscible or difficult to mix with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2362-10-9
MDL Number MFCD00000877

Specifications

Boiling Point 92°C to 93°C (20mmHg)
Quantity 50 g
Molecular Formula C7H7BrO
Beilstein 506631
Flash Point 73°C (163°F)
Synonym 4-bromo-o-cresol, 2-methyl-4-bromophenol, 4-bromo-2-methyl-phenol, o-cresol, 4-bromo, phenol, 4-bromo-2-methyl, 2-hydroxy-5-bromotoluene, p-bromocresol, 4-bromo-cresol, pubchem7106, acmc-1chy8
Solubility Information Not miscible or difficult to mix with water.
InChI Key IWJGMJHAIUBWKT-UHFFFAOYSA-N
SMILES CC1=C(C=CC(=C1)Br)O
IUPAC Name 4-bromo-2-methylphenol
Molecular Weight (g/mol) 187.04
PubChem CID 16896
Formula Weight 231.27
Percent Purity 97%
Refractive Index 1.44
Density 1.05
Chemical Name or Material 1,3-Bis(chloromethyl)tetramethyldisiloxane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'