missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,10-Phenanthroline Hydrochloride Monohydrate,99.5 to 102% (calc. on dry substance), Honeywell™ Fluka™
Reagent European Pharmacopoeia, for spectrophotometric det., 99.5-102% (calc. on dry substance)
£57.88 - £1021.24
Chemical Identifiers
CAS | 18851-33-7 |
---|---|
Molecular Formula | C12H11ClN2O |
Molecular Weight (g/mol) | 234.683 |
MDL Number | MFCD00150061 |
InChI Key | NDLHUHRGAIHALB-UHFFFAOYSA-N |
Synonym | 1,10-phenanthroline hydrochloride monohydrate, 1,10-phenanthroline hydrochloride hydrate, 1,10-phenanthroline monohydrochloride monohydrate, 1,10-phenanthroline hydrate hydrochloride, 1,10-phenanthroline, monohydrochloride, monohydrate, phen hydrate hydrochloride, acmc-209iz9, dsstox_cid_24309, dsstox_rid_80145, dsstox_gsid_44309 |
PubChem CID | 2723715 |
IUPAC Name | 1,10-phenanthroline;hydrate;hydrochloride |
SMILES | C1=CC2=C(C3=C(C=CC=N3)C=C2)N=C1.O.Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15683170
|
Honeywell-Fluka
31043-5G |
5 g | Glass Bottle |
£66.88
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15663170
|
Honeywell-Fluka
31043-10G |
10 g | Glass Bottle |
£57.88
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15673170
|
Honeywell-Fluka
31043-500G |
500 g | Glass Bottle | N/A | |||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
18851-33-7 | |
234.683 | |
NDLHUHRGAIHALB-UHFFFAOYSA-N | |
2723715 | |
C1=CC2=C(C3=C(C=CC=N3)C=C2)N=C1.O.Cl |
C12H11ClN2O | |
MFCD00150061 | |
1,10-phenanthroline hydrochloride monohydrate, 1,10-phenanthroline hydrochloride hydrate, 1,10-phenanthroline monohydrochloride monohydrate, 1,10-phenanthroline hydrate hydrochloride, 1,10-phenanthroline, monohydrochloride, monohydrate, phen hydrate hydrochloride, acmc-209iz9, dsstox_cid_24309, dsstox_rid_80145, dsstox_gsid_44309 | |
1,10-phenanthroline;hydrate;hydrochloride |
Specifications
18851-33-7 | |
C12H11ClN2O | |
UN2811 | |
1,10-phenanthroline hydrochloride monohydrate, 1,10-phenanthroline hydrochloride hydrate, 1,10-phenanthroline monohydrochloride monohydrate, 1,10-phenanthroline hydrate hydrochloride, 1,10-phenanthroline, monohydrochloride, monohydrate, phen hydrate hydrochloride, acmc-209iz9, dsstox_cid_24309, dsstox_rid_80145, dsstox_gsid_44309 | |
C1=CC2=C(C3=C(C=CC=N3)C=C2)N=C1.O.Cl | |
234.683 | |
234.68g/mol | |
1,10-Phenanthroline Hydrochloride Monohydrate, Reag. Ph. Eur. |
224°C to 225°C | |
MFCD00150061 | |
4007967 | |
NDLHUHRGAIHALB-UHFFFAOYSA-N | |
1,10-phenanthroline;hydrate;hydrochloride | |
2723715 | |
99.5 to 102% (calc. on dry substance) |
Safety and Handling
P273-P301 + P310-P501
H301-H410
missing translation for 'dotInformation' : Class 6.1 - PG 3 - EHS - Toxic solids, organic, n.o.s., HI: all
missing translation for 'einecsNumber' : 223-325-1
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- 1,10-Phenanthroline hydrochloride monohydrate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
Hazard Statement
- H301-Toxic if swallowed.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : 1,10-Phenanthrolinium chloride monohydrate
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title