missing translation for 'onlineSavingsMsg'
Learn More

1,10-Phenanthroline Hydrochloride Monohydrate,99.5 to 102% (calc. on dry substance), Honeywell™ Fluka™

Product Code. 15663170 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
5 g
10 g
500 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Reagent European Pharmacopoeia, for spectrophotometric det., 99.5-102% (calc. on dry substance)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18851-33-7
Molecular Formula C12H11ClN2O
Molecular Weight (g/mol) 234.683
MDL Number MFCD00150061
InChI Key NDLHUHRGAIHALB-UHFFFAOYSA-N
Synonym 1,10-phenanthroline hydrochloride monohydrate, 1,10-phenanthroline hydrochloride hydrate, 1,10-phenanthroline monohydrochloride monohydrate, 1,10-phenanthroline hydrate hydrochloride, 1,10-phenanthroline, monohydrochloride, monohydrate, phen hydrate hydrochloride, acmc-209iz9, dsstox_cid_24309, dsstox_rid_80145, dsstox_gsid_44309
PubChem CID 2723715
IUPAC Name 1,10-phenanthroline;hydrate;hydrochloride
SMILES C1=CC2=C(C3=C(C=CC=N3)C=C2)N=C1.O.Cl

Specifications

Melting Point 224°C to 225°C
Packaging Glass Bottle
Quantity 10 g
UN Number UN2811
Beilstein 4007967
Formula Weight 234.68g/mol
Percent Purity 99.5 to 102% (calc. on dry substance)
Chemical Name or Material 1,10-Phenanthroline Hydrochloride Monohydrate, Reag. Ph. Eur.
compliance-icons compliance-icons
Product Identifier
  • 1,10-Phenanthroline hydrochloride monohydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 1,10-Phenanthrolinium chloride monohydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.