Learn More
Tris(3,6-dioxaheptyl)amine, 95%, Thermo Scientific Chemicals
£21.50 - £99.40
Chemical Identifiers
CAS | 70384-51-9 |
---|---|
Molecular Formula | C15H33NO6 |
Molecular Weight (g/mol) | 323.43 |
MDL Number | MFCD00010748 |
InChI Key | XGLVDUUYFKXKPL-UHFFFAOYSA-N |
Synonym | tris 2-2-methoxyethoxy ethyl amine, tris dioxa-3,6-heptyl amine, tris 3,6-dioxaheptyl amine, tda-1, 2-2-methoxyethoxy-n,n-bis 2-2-methoxyethoxy ethyl ethanamine, ethanamine, 2-2-methoxyethoxy-n,n-bis 2-2-methoxyethoxy ethyl, 8-2-2-methoxyethoxy ethyl-2,5,11,14-tetraoxa-8-azapentadecane, tris-2-2-methoxyethoxy ethyl amine |
PubChem CID | 112414 |
IUPAC Name | 2-(2-methoxyethoxy)-N,N-bis[2-(2-methoxyethoxy)ethyl]ethanamine |
SMILES | COCCOCCN(CCOCCOC)CCOCCOC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11313137
|
Thermo Scientific Alfa Aesar
L13544.06 |
5 g |
£21.50
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11323137
|
Thermo Scientific Alfa Aesar
L13544.14 |
25 g |
£32.50
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11333137
|
Thermo Scientific Alfa Aesar
L13544.22 |
100 g |
£99.40
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Tris(3,6-dioxaheptyl)amine acts as a phase transfer catalyst used in the preparation of 1,3-dideaza-2'-deoxyadenosine and related benzimidazole 2'-deoxyribonucleosides. It is also used as an intermediate in the synthesis of telithromycin. Further, it is used in the oxidation of arylmethanols under phase-transfer conditions. In addition to this, it acts as a cryptand during phase-transfer glycosylation of 2-amino-4-methoxy-7H-pyrrolo[2,3-d]pyrimidine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTris(3,6-dioxaheptyl)amine acts as a phase transfer catalyst used in the preparation of 1,3-dideaza-2′-deoxyadenosine and related benzimidazole 2′-deoxyribonucleosides. It is also used as an intermediate in the synthesis of telithromycin. Further, it is used in the oxidation of arylmethanols under phase-transfer conditions. In addition to this, it acts as a cryptand during phase-transfer glycosylation of 2-amino-4-methoxy-7H-pyrrolo[2,3-d]pyrimidine.
Solubility
Miscible with water.
Notes
Air sensitive and hygroscopic. Incompatible strong oxidizing agents and strong acids.
Chemical Identifiers
70384-51-9 | |
323.43 | |
XGLVDUUYFKXKPL-UHFFFAOYSA-N | |
112414 | |
COCCOCCN(CCOCCOC)CCOCCOC |
C15H33NO6 | |
MFCD00010748 | |
tris 2-2-methoxyethoxy ethyl amine, tris dioxa-3,6-heptyl amine, tris 3,6-dioxaheptyl amine, tda-1, 2-2-methoxyethoxy-n,n-bis 2-2-methoxyethoxy ethyl ethanamine, ethanamine, 2-2-methoxyethoxy-n,n-bis 2-2-methoxyethoxy ethyl, 8-2-2-methoxyethoxy ethyl-2,5,11,14-tetraoxa-8-azapentadecane, tris-2-2-methoxyethoxy ethyl amine | |
2-(2-methoxyethoxy)-N,N-bis[2-(2-methoxyethoxy)ethyl]ethanamine |
Specifications
70384-51-9 | |
1.011 | |
162°C (323°F) | |
1.4486 | |
2369296 | |
tris 2-2-methoxyethoxy ethyl amine, tris dioxa-3,6-heptyl amine, tris 3,6-dioxaheptyl amine, tda-1, 2-2-methoxyethoxy-n,n-bis 2-2-methoxyethoxy ethyl ethanamine, ethanamine, 2-2-methoxyethoxy-n,n-bis 2-2-methoxyethoxy ethyl, 8-2-2-methoxyethoxy ethyl-2,5,11,14-tetraoxa-8-azapentadecane, tris-2-2-methoxyethoxy ethyl amine | |
XGLVDUUYFKXKPL-UHFFFAOYSA-N | |
2-(2-methoxyethoxy)-N,N-bis[2-(2-methoxyethoxy)ethyl]ethanamine | |
112414 | |
95% |
<-100°C | |
163°C to 171°C (1 mmHg) | |
C15H33NO6 | |
MFCD00010748 | |
Air sensitive; Hygroscopic | |
Miscible with water. | |
COCCOCCN(CCOCCOC)CCOCCOC | |
323.43 | |
323.43 | |
Tris(3,6-dioxaheptyl)amine |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: III
missing translation for 'einecsNumber' : 274-590-5
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only