Learn More
trans-2,5-Dimethylpiperazine, 98%, Thermo Scientific Chemicals
£29.70 - £345.00
Chemical Identifiers
CAS | 2815-34-1 |
---|---|
Molecular Formula | C6H14N2 |
Molecular Weight (g/mol) | 114.192 |
MDL Number | MFCD00070516 |
InChI Key | NSMWYRLQHIXVAP-OLQVQODUSA-N |
Synonym | trans-2,5-dimethylpiperazine, 2r,5s-2,5-dimethylpiperazine, 2s,5r-2,5-dimethylpiperazine, 2s,5r-2,5-dimethyl-piperazine, piperazine, 2,5-dimethyl-, 2r,5s-rel, 2r,5s-2,5-dimethylpiperazin, piperazine, 2,5-dimethyl-, trans, 2r,5s-2,5-dimethyl-piperazine, pubchem15878, 2,5-dimethylpiperazine # |
PubChem CID | 220672 |
IUPAC Name | (2R,5S)-2,5-dimethylpiperazine |
SMILES | CC1CNC(CN1)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11486376
|
Thermo Scientific Alfa Aesar
B23150.06 |
5 g |
£29.70
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11496376
|
Thermo Scientific Alfa Aesar
B23150.14 |
25 g |
£94.20
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11406386
|
Thermo Scientific Alfa Aesar
B23150.22 |
100 g |
£345.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2815-34-1 | |
114.192 | |
NSMWYRLQHIXVAP-OLQVQODUSA-N | |
220672 | |
CC1CNC(CN1)C |
C6H14N2 | |
MFCD00070516 | |
trans-2,5-dimethylpiperazine, 2r,5s-2,5-dimethylpiperazine, 2s,5r-2,5-dimethylpiperazine, 2s,5r-2,5-dimethyl-piperazine, piperazine, 2,5-dimethyl-, 2r,5s-rel, 2r,5s-2,5-dimethylpiperazin, piperazine, 2,5-dimethyl-, trans, 2r,5s-2,5-dimethyl-piperazine, pubchem15878, 2,5-dimethylpiperazine # | |
(2R,5S)-2,5-dimethylpiperazine |
Specifications
2815-34-1 | |
162°C to 165°C | |
C6H14N2 | |
UN2925 | |
trans-2,5-dimethylpiperazine, 2r,5s-2,5-dimethylpiperazine, 2s,5r-2,5-dimethylpiperazine, 2s,5r-2,5-dimethyl-piperazine, piperazine, 2,5-dimethyl-, 2r,5s-rel, 2r,5s-2,5-dimethylpiperazin, piperazine, 2,5-dimethyl-, trans, 2r,5s-2,5-dimethyl-piperazine, pubchem15878, 2,5-dimethylpiperazine # | |
CC1CNC(CN1)C | |
114.192 | |
114.19 | |
trans-2,5-Dimethylpiperazine |
115°C to 119°C | |
58°C (136°F) | |
MFCD00070516 | |
79886 | |
NSMWYRLQHIXVAP-OLQVQODUSA-N | |
(2R,5S)-2,5-dimethylpiperazine | |
220672 | |
98% |
Safety and Handling
GHS H Statement
H314-H318-H228-H312
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable solid.
Harmful in contact with skin.
P210-P240-P241-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H228-H312-H314-H500
missing translation for 'dotInformation' : Hazard Class: 4.1; Packaging Group: II
missing translation for 'einecsNumber' : 000-000-0
missing translation for 'rtecsNumber' : TL5950000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only