missing translation for 'onlineSavingsMsg'
Learn More

trans-2,5-Dimethylpiperazine, 98%, Thermo Scientific Chemicals

Product Code. 11486376
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2815-34-1
Molecular Formula C6H14N2
Molecular Weight (g/mol) 114.192
MDL Number MFCD00070516
InChI Key NSMWYRLQHIXVAP-OLQVQODUSA-N
Synonym trans-2,5-dimethylpiperazine, 2r,5s-2,5-dimethylpiperazine, 2s,5r-2,5-dimethylpiperazine, 2s,5r-2,5-dimethyl-piperazine, piperazine, 2,5-dimethyl-, 2r,5s-rel, 2r,5s-2,5-dimethylpiperazin, piperazine, 2,5-dimethyl-, trans, 2r,5s-2,5-dimethyl-piperazine, pubchem15878, 2,5-dimethylpiperazine #
PubChem CID 220672
IUPAC Name (2R,5S)-2,5-dimethylpiperazine
SMILES CC1CNC(CN1)C

Specifications

Melting Point 115°C to 119°C
Boiling Point 162°C to 165°C
Flash Point 58°C (136°F)
Quantity 5 g
UN Number UN2925
Beilstein 79886
Formula Weight 114.19
Percent Purity 98%
Chemical Name or Material trans-2,5-Dimethylpiperazine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'