missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyldimethylsilylacetylene, 98%, Thermo Scientific Chemicals

£56.80 - £707.00

Chemical Identifiers

CAS 86318-61-8
Molecular Formula C8H16Si
Molecular Weight (g/mol) 140.301
MDL Number MFCD00191877
InChI Key RTYNRTUKJVYEIE-UHFFFAOYSA-N
Synonym tert-butyldimethylsilyl acetylene, tert-butyldimethylsilylacetylene, tert-butyl ethynyl dimethylsilane, ethynyl-t-butyldimethylsilane, tert-butyldimethylsilyl ethyne, tert-butyl-ethynyl-dimethyl-silane, silane, 1,1-dimethylethyl ethynyldimethyl, t-butyldimethylsilylacetylene, tert-butyldimethylsilylethyne, tert-butylethynyldimethylsilane
PubChem CID 2757281
IUPAC Name tert-butyl-ethynyl-dimethylsilane
SMILES CC(C)(C)[Si](C)(C)C#C
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15440378
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H53494.03
1 g
£56.80
1g
Please to purchase this item. Need a web account? Register with us today!
15450378
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H53494.06
5 g
£195.00
5g
Please to purchase this item. Need a web account? Register with us today!
15460378
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H53494.14
25 g
£707.00
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

tert-Butyldimethylsilylacetylene may be used in the synthesis of β-alkynylketone and β-alkynyl aldehydes. It participates in Cadiot-Chodkiewicz cross-coupling reaction with various bromoalkynes to afford synthetically useful unsymmetrical diynes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyldimethylsilylacetylene may be used in the synthesis of β-alkynylketone and β-alkynyl aldehydes. It participates in Cadiot-Chodkiewicz cross-coupling reaction with various bromoalkynes to afford synthetically useful unsymmetrical diynes.

Solubility
Sparingly soluble in water (0.043 g/L at 25°C).

Notes
Stable under recommended storage conditions.
Specifications

Chemical Identifiers

86318-61-8
140.301
RTYNRTUKJVYEIE-UHFFFAOYSA-N
2757281
CC(C)(C)[Si](C)(C)C#C
C8H16Si
MFCD00191877
tert-butyldimethylsilyl acetylene, tert-butyldimethylsilylacetylene, tert-butyl ethynyl dimethylsilane, ethynyl-t-butyldimethylsilane, tert-butyldimethylsilyl ethyne, tert-butyl-ethynyl-dimethyl-silane, silane, 1,1-dimethylethyl ethynyldimethyl, t-butyldimethylsilylacetylene, tert-butyldimethylsilylethyne, tert-butylethynyldimethylsilane
tert-butyl-ethynyl-dimethylsilane

Specifications

130°C
C8H16Si
MFCD00191877
1921466
tert-butyldimethylsilyl acetylene, tert-butyldimethylsilylacetylene, tert-butyl ethynyl dimethylsilane, ethynyl-t-butyldimethylsilane, tert-butyldimethylsilyl ethyne, tert-butyl-ethynyl-dimethyl-silane, silane, 1,1-dimethylethyl ethynyldimethyl, t-butyldimethylsilylacetylene, tert-butyldimethylsilylethyne, tert-butylethynyldimethylsilane
RTYNRTUKJVYEIE-UHFFFAOYSA-N
tert-butyl-ethynyl-dimethylsilane
2757281
98%
0.751
86318-61-8
(CH3)3CSi(CH3)2C≡CH
UN1993
6°C (43°F)
Sparingly soluble in water (0.043g/L at 25°C).
CC(C)(C)[Si](C)(C)C#C
140.301
140.3
1.451
tert-Butyldimethylsilylacetylene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H225-H315-H319-H335

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only