missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Butyldimethylsilylacetylene, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
tert-Butyldimethylsilylacetylene may be used in the synthesis of β-alkynylketone and β-alkynyl aldehydes. It participates in Cadiot-Chodkiewicz cross-coupling reaction with various bromoalkynes to afford synthetically useful unsymmetrical diynes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
tert-Butyldimethylsilylacetylene may be used in the synthesis of β-alkynylketone and β-alkynyl aldehydes. It participates in Cadiot-Chodkiewicz cross-coupling reaction with various bromoalkynes to afford synthetically useful unsymmetrical diynes.
Solubility
Sparingly soluble in water (0.043 g/L at 25°C).
Notes
Stable under recommended storage conditions.
tert-Butyldimethylsilylacetylene may be used in the synthesis of β-alkynylketone and β-alkynyl aldehydes. It participates in Cadiot-Chodkiewicz cross-coupling reaction with various bromoalkynes to afford synthetically useful unsymmetrical diynes.
Solubility
Sparingly soluble in water (0.043 g/L at 25°C).
Notes
Stable under recommended storage conditions.
Specifications
Specifications
Boiling Point | 130°C |
Quantity | 5 g |
Linear Formula | (CH3)3CSi(CH3)2C≡CH |
UN Number | UN1993 |
Beilstein | 1921466 |
Flash Point | 6°C (43°F) |
Solubility Information | Sparingly soluble in water (0.043g/L at 25°C). |
Formula Weight | 140.3 |
Percent Purity | 98% |
Refractive Index | 1.451 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'