Learn More
(S)-(+)-2,3-O-Isopropylideneglycerol, 98%, Thermo Scientific Chemicals
£28.50 - £80.00
Chemical Identifiers
CAS | 22323-82-6 |
---|---|
Molecular Formula | C6H12O3 |
Molecular Weight (g/mol) | 132.16 |
MDL Number | MFCD00063239 |
InChI Key | RNVYQYLELCKWAN-UHFFFAOYNA-N |
Synonym | s-+-2,2-dimethyl-1,3-dioxolane-4-methanol, s-glycerol acetonide, s-+-1,2-isopropylideneglycerol, 4s-2,2-dimethyl-1,3-dioxolan-4-yl methanol, s-2,2-dimethyl-1,3-dioxolane-4-methanol, s-solketal, 1,2-isopropylidene-sn-glycerol, s-+-2,3-o-isopropylideneglycerol, 1,3-dioxolane-4-methanol, 2,2-dimethyl-, 4s, s-2,2-dimethyl-1,3-dioxolan-4-yl methanol |
PubChem CID | 736057 |
IUPAC Name | [(4S)-2,2-dimethyl-1,3-dioxolan-4-yl]methanol |
SMILES | CC1(C)OCC(CO)O1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11415957
|
Thermo Scientific Alfa Aesar
B24488.04 |
2 g |
£28.50
2g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11425957
|
Thermo Scientific Alfa Aesar
B24488.09 |
10 g |
£80.00
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
(S)-(+)-2,3-O-Isopropylideneglycerol acts as a MEK inhibitor with antitumor activity. It is also employed in the preparation of a chiral allylic triol through extrusion of sulfur dioxide from an alfa,beta-epoxysulfone.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(S)-(+)-2,3-O-Isopropylideneglycerol acts as a MEK inhibitor with antitumor activity. It is also employed in the preparation of a chiral allylic triol through extrusion of sulfur dioxide from an alfa,beta-epoxysulfone.
Solubility
Miscible with water and chloroform.
Notes
Store in a cool place. Incompatible with acids and strong oxidizing agents.
Chemical Identifiers
22323-82-6 | |
132.16 | |
RNVYQYLELCKWAN-UHFFFAOYNA-N | |
736057 | |
CC1(C)OCC(CO)O1 |
C6H12O3 | |
MFCD00063239 | |
s-+-2,2-dimethyl-1,3-dioxolane-4-methanol, s-glycerol acetonide, s-+-1,2-isopropylideneglycerol, 4s-2,2-dimethyl-1,3-dioxolan-4-yl methanol, s-2,2-dimethyl-1,3-dioxolane-4-methanol, s-solketal, 1,2-isopropylidene-sn-glycerol, s-+-2,3-o-isopropylideneglycerol, 1,3-dioxolane-4-methanol, 2,2-dimethyl-, 4s, s-2,2-dimethyl-1,3-dioxolan-4-yl methanol | |
[(4S)-2,2-dimethyl-1,3-dioxolan-4-yl]methanol |
Specifications
22323-82-6 | |
82°C to 83°C (14 mmHg) | |
C6H12O3 | |
MFCD00063239 | |
14,5213 | |
Miscible with water and chloroform. | |
+11.5° (c=5 in Methanol) | |
[(4S)-2,2-dimethyl-1,3-dioxolan-4-yl]methanol | |
736057 | |
98% |
1.066 | |
80°C (176°F) | |
1.435 | |
80118 | |
s-+-2,2-dimethyl-1,3-dioxolane-4-methanol, s-glycerol acetonide, s-+-1,2-isopropylideneglycerol, 4s-2,2-dimethyl-1,3-dioxolan-4-yl methanol, s-2,2-dimethyl-1,3-dioxolane-4-methanol, s-solketal, 1,2-isopropylidene-sn-glycerol, s-+-2,3-o-isopropylideneglycerol, 1,3-dioxolane-4-methanol, 2,2-dimethyl-, 4s, s-2,2-dimethyl-1,3-dioxolan-4-yl methanol | |
RNVYQYLELCKWAN-UHFFFAOYNA-N | |
CC1(C)OCC(CO)O1 | |
132.16 | |
132.18 | |
(S)-(+)-2,3-O-Isopropylideneglycerol |
Safety and Handling
GHS H Statement
H227
Combustible liquid.
P210-P280-P370+P378q-P501c
H227
missing translation for 'einecsNumber' : 244-910-8
missing translation for 'tsca' : No
Recommended Storage : Keep cold
RUO – Research Use Only