missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-(+)-2,3-O-Isopropylideneglycerol, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
2 g
10 g
Description
(S)-(+)-2,3-O-Isopropylideneglycerol acts as a MEK inhibitor with antitumor activity. It is also employed in the preparation of a chiral allylic triol through extrusion of sulfur dioxide from an alfa,beta-epoxysulfone.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(S)-(+)-2,3-O-Isopropylideneglycerol acts as a MEK inhibitor with antitumor activity. It is also employed in the preparation of a chiral allylic triol through extrusion of sulfur dioxide from an alfa,beta-epoxysulfone.
Solubility
Miscible with water and chloroform.
Notes
Store in a cool place. Incompatible with acids and strong oxidizing agents.
(S)-(+)-2,3-O-Isopropylideneglycerol acts as a MEK inhibitor with antitumor activity. It is also employed in the preparation of a chiral allylic triol through extrusion of sulfur dioxide from an alfa,beta-epoxysulfone.
Solubility
Miscible with water and chloroform.
Notes
Store in a cool place. Incompatible with acids and strong oxidizing agents.
Specifications
Specifications
Density | 1.066 |
Boiling Point | 82°C to 83°C (14 mmHg) |
Flash Point | 80°C (176°F) |
Refractive Index | 1.435 |
Quantity | 2 g |
Beilstein | 80118 |
Merck Index | 14,5213 |
Solubility Information | Miscible with water and chloroform. |
Optical Rotation | +11.5° (c=5 in Methanol) |
Formula Weight | 132.18 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'