missing translation for 'onlineSavingsMsg'
Learn More

Restek™ Oil-Free Vacuum/Pressure Station for Resprep Systems

For 12-Port Resprep™ SPE Systems.

Brand:  Restek™ 24002

Code : Z3

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 13149922

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

  • Allows switching from pressure to vacuum in a matter of seconds
  • Quiet, oil-free unit will not contaminate the extraction system or samples
  • Provides 20 in. Hg (68 kPa) vacuum or 18psi (124 kPa) pressure

Order Info

Not recommended for use with 24-port manifold.

Specifications

Specifications

To Allow Switching From Pressure to Vacuum
Oil-free Vacuum/Pressure Station
115 V 60 Hz
60 Hz
12 Port Resprep SPE System
18 psi
115 V
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers