Learn More
JG Finneran Associates™ PTFE Seals
Use these inserts with dram sample vials. JG Finneran™ dram sample vials are made of Type I borosilicate glass, providing exceptional resistance to heat shock and chemical leaching.
Brand:  JG Finneran Associates™ 601010-13
Code : 17
Additional Details : Weight : 0.08100kg
 View more versions of this product
Product Code. 10187500
  • £106.30 / Pack of 1000
Estimated Shipment: 21-05-2024
to see stock.
Add to basket
Description
Description
  • PTFE seals
  • For use with dram sample vials
  • Available in a variety of sizes
Specifications
Specifications
PTFE
White
1000 Pk.
13 mm