missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Protein Kinase Inhibitor

£160.00 - £378.00

Chemical Identifiers

CAS 97161-97-2
Molecular Formula C27H21N3O5
Synonym K252a
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15424239
View Documents Promotion Details
Thermo Scientific Chemicals
J61687.M^
1 mg
£160.00
1mg
Please to purchase this item. Need a web account? Register with us today!
15414239
View Documents Promotion Details
Thermo Scientific Chemicals
J61687.#0
5 mg
£378.00
5mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Inhibitor of protein kinase A

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

97161-97-2
C27H21N3O5
K252a

Specifications

Off-white
Inhibitor of protein kinase A
C27H21N3O5
K252a
KOZFSFOOLUUIGY-UHFFFAOYNA-N
methyl 16-hydroxy-15-methyl-3-oxo-28-oxa-4,14,19-triazaoctacyclo[12.11.2.1¹⁵,¹⁸.0²,⁶.0⁷,²⁷.0⁸,¹³.0¹⁹,²⁶.0²⁰,²⁵]octacosa-1(26),2(6),7(27),8(13),9,11,20,22,24-nonaene-16-carboxylate
-30°C to -10°C
97161-97-2
467.47
Powder
COC(=O)C1(O)CC2OC1(C)N1C3=C(C=CC=C3)C3=C1C1=C(C4=CC=CC=C4N21)C1=C3CNC1=O
467.48
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only