missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Protein Kinase Inhibitor

Product Code. 15414239
Click to view available options
Quantity:
1 mg
5 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Inhibitor of protein kinase A

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97161-97-2
Molecular Formula C27H21N3O5
Synonym K252a

Specifications

Quantity 5 mg
Description Off-white
Content And Storage -30°C to -10°C
For Use With (Application) Inhibitor of protein kinase A
Formula Weight 467.47
Form Powder
InChI Key KOZFSFOOLUUIGY-UHFFFAOYNA-N
SMILES COC(=O)C1(O)CC2OC1(C)N1C3=C(C=CC=C3)C3=C1C1=C(C4=CC=CC=C4N21)C1=C3CNC1=O
IUPAC Name methyl 16-hydroxy-15-methyl-3-oxo-28-oxa-4,14,19-triazaoctacyclo[12.11.2.1¹⁵,¹⁸.0²,⁶.0⁷,²⁷.0⁸,¹³.0¹⁹,²⁶.0²⁰,²⁵]octacosa-1(26),2(6),7(27),8(13),9,11,20,22,24-nonaene-16-carboxylate
Molecular Weight (g/mol) 467.48

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'