missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VITLAB™ VITsafe™ 500mL Wide-Mouth Wash Bottles
Click to view available options
Material:
LDPE
Description:
Imprint: Acetone
Imprint: Acetonitrile
Imprint: Distilled Water
Imprint: Ethanol
Imprint: Ethyl acetate
Imprint: Methanol
Imprint: Methylene chloride
Imprinted: Isopropanol
Imprinted: N,N-Dimethylformamide
Imprinted: Toluene
Closure Color:
Blue
Green
Red
Yellow
Description
- Manufactured from PE-LD (or PP for Acetone and MEK), spray tube from PP
- Complies in accordance to Directive (EC) No. 1272/2008 (GHS)
- Imprinted with:
- Material name in German, English, French and Spanish
- Chemical formula, CAS No, hazard pictogram, signal word
- Risk Phrases (H phrases), safety phrases (P phrases), as well as NFPA Code
- Precise spray jet and optimised medium backflow through the continuous and finely drawn tip of the spray tube
- The VENT-CAP design oprevents almost all static overpressure minimizing leakage or dripping
Specifications
Capacity (Metric) | 500 mL |
Description | Imprint: Distilled Water |
Material | LDPE |
Closure Color | Blue |
Certifications/Compliance | Directive (EC) No. 1272/2008 (GHS) |
Closure Included | Yes |
Mouth | Wide |
Height (Metric) | 166 mm |
Closure Material | Polypropylene |
Diameter (Metric) | 76 mm |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'