missing translation for 'onlineSavingsMsg'
Learn More

VITLAB™ VITsafe™ 500mL Wide-Mouth Wash Bottles

Codice prodotto. 15792056
Click to view available options
Material:
LDPE
Description:
Imprint: Acetone
Imprint: Acetonitrile
Imprint: Distilled Water
Imprint: Ethanol
Imprint: Ethyl acetate
Imprint: Methanol
Imprint: Methylene chloride
Imprinted: Isopropanol
Imprinted: N,N-Dimethylformamide
Imprinted: Toluene
Closure Color:
Blue
Green
Red
Yellow
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Imprinted with durable safety symbols, and other important information for optimum laboratory safety. Vitlab™ VITsafe™ Wide-Mouth Wash Bottles include color coded vent-cap that supports easy visual identification of the media.

  • Manufactured from PE-LD (or PP for Acetone and MEK), spray tube from PP
  • Complies in accordance to Directive (EC) No. 1272/2008 (GHS)
  • Imprinted with:
    • Material name in German, English, French and Spanish
    • Chemical formula, CAS No, hazard pictogram, signal word
    • Risk Phrases (H phrases), safety phrases (P phrases), as well as NFPA Code
  • Precise spray jet and optimised medium backflow through the continuous and finely drawn tip of the spray tube
  • The VENT-CAP design oprevents almost all static overpressure minimizing leakage or dripping

Specifica

Capacity (Metric) 500 mL
Description Imprint: Methylene chloride
Includes Screw Cap, Spray Tube
Material LDPE
Closure Color Red
Certifications/Compliance Directive (EC) No. 1272/2008 (GHS)
Closure Included Yes
Mouth Wide
Height (Metric) 166 mm
Closure Material Polypropylene
Diameter (Metric) 76 mm
Closure Type Vented
Vedi altri risultati Mostra meno risultati
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'