missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Biodesign™ Nylon Matrix Filter CellMicroSieves™
Allow the rapid, yet gentle, isolation of cells, organelles, or cellular debris.
Brand: Biodesign™ N5S
Code : LB
Additional Details : Weight : 0.10000kg
Description
Many biological preparations begin with extractions that require coarse filtration to remove cellular debris. Bulk filtration using cheesecloth absorbs much of the extraction fluid, while membranes clog quickly. BioPrepNylon™ produces rapid bulk filtration at extraordinary flow rates. Polypropylene sieve holders (2 piece) holders with an adjustable metal tightener hold BioPrepNylon™ securely when processing samples. BioPrepNylon™ is loosely placed in a sieve holder to form a funnel. The sample is placed in the funnel, allowed to drain, and the layers of the BioPrepNylon™ are gently pressed together to collect all of the extraction liquid.- A pure nylon product without detergents, dyes or salts, can be used directly with biological samples
- Consists of very fine nylon filaments woven into a matrix with 50μm pores
- As BioPrepNylon™ does not wet, significantly greater quantities of the extraction liquid are recovered compared to any other method
- BioPrepNylon™ does not bind proteins, so no loss of your valuable sample will occur
- Compatible with the salts, buffers and solvents used in biological research
- Autoclavable
- High tensile strength
- Can be repeatedly washed and reused BioPrepNylon™ is a synthetic cheese cloth for rapid bulk filtration.
- A pure nylon product that is biologically inert, resistant to most acidic or basic conditions and many solvents
- Autoclavable and can be repeatedly washed and reused
- Rapid collection of tissue culture cells without centrifugation
- Isolation of specific cell types from a dissociated tissue sample, allowing only a single type of cell to pass through
- Separation of different sized cells from a mixture of cell types
- Concentration and isolation of cells from a bioreactor
- Prefilter to prevent clogging of final filter CellMicroSieves™ allow the rapid gentle isolation of cells, organelles or cellular debris.
Specifications
For rapid collection of tissue culture cells without centrifugation | |
3Pack | |
Matrix Filter CellMicroSieves |
Yes | |
5μm | |
Nylon |