missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Biodesign™ Nylon Matrix Filter CellMicroSieves™
Allow the rapid, yet gentle, isolation of cells, organelles, or cellular debris.
£57.45 - £237.00
Specifications
Autoclavable | Yes |
---|---|
For Use With (Application) | For rapid collection of tissue culture cells without centrifugation |
Type | Matrix Filter CellMicroSieves |
Material | Nylon |
Product Code | Brand | Pore Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | Price | Quantity & Availability | |||||
12974257
|
Biodesign™
N10S |
10μm |
£237.00
Pack of 3 |
Estimated Shipment: 15-05-2024
Log in to see stock. |
|||||
12684787
|
Biodesign™
N100S |
100μm |
£91.30
Pack of 3 |
Estimated Shipment: 15-05-2024
Log in to see stock. |
|||||
12914277
|
Biodesign™
N200C |
200μm |
£80.55
Pack of 10 |
Estimated Shipment: 15-05-2024
Log in to see stock. |
|||||
12964257
|
Biodesign™
N15S |
15μm |
£75.30
Pack of 3 |
Estimated Shipment: 20-05-2024
Log in to see stock. |
|||||
12904267
|
Biodesign™
N200R |
200μm |
£122.00
Each |
Estimated Shipment: 15-05-2024
Log in to see stock. |
|||||
12994247
|
Biodesign™
N200S |
200μm |
£148.00
Pack of 3 |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
12954257
|
Biodesign™
N20S |
20μm |
£131.00
Pack of 3 |
Estimated Shipment: 15-05-2024
Log in to see stock. |
|||||
12944257
|
Biodesign™
N25S |
25μm |
£137.00
Pack of 3 |
Estimated Shipment: 15-05-2024
Log in to see stock. |
|||||
12987487
|
Biodesign™
N305 |
30μm |
£104.00
Each |
Estimated Shipment: 18-07-2024
Log in to see stock. |
|||||
12944267
|
Biodesign™
N35R |
35μm |
£122.00
Each |
Estimated Shipment: 18-07-2024
Log in to see stock. |
|||||
12924257
|
Biodesign™
N50S |
50μm |
£80.35
Pack of 3 |
Estimated Shipment: 15-05-2024
Log in to see stock. |
|||||
12904277
|
Biodesign™
N5R |
5μm |
£185.00
Each |
Estimated Shipment: 15-05-2024
Log in to see stock. |
|||||
12994257
|
Biodesign™
N5S |
5μm |
£101.00
Pack of 3 |
Estimated Shipment: 15-05-2024
Log in to see stock. |
|||||
12914257
|
Biodesign™
N70S |
70μm |
£57.45
Pack of 3 |
Estimated Shipment: 18-07-2024
Log in to see stock. |
|||||
Description
Unlike traditional filters, CellMicroSieves™ are a precision woven matrix of pure nylon filaments that is available with pores ranging from 5μL to 200μL. CellMicroSieves™ do not wet and contains no dyes, detergents or extractables. Extremely low protein affinity means that cells and proteins will not bind to CellMicroSieves™. They have a wide variety of distinctive uses a specific sized porous screens in the laboratory. AccessoriesPolypropylene sieve holders (2 piece) holders with adjustable metal tightener or filtration units, type A borosilicate glass with stainless steel filter support and aluminum clamp or type B complete polysulfone filtration unit with polypropylene filter support are available. Both filtration units have a 300mL capacity are autoclavable and reusable. Use sheets and rolls with sieve holders and circles with filtration units to hold securely when processing samples.- A pure nylon product that is biologically inert, resistant to most acidic or basic conditions and many solvents
- Autoclavable and can be repeatedly washed and reused
- Rapid collection of tissue culture cells without centrifugation
- Isolation of specific cell types from a dissociated tissue sample, allowing only a single type of cell to pass through
- Separation of different sized cells from a mixture of cell types
- Concentration and isolation of cells from a bioreactor
- Prefilter to prevent clogging of final filter CellMicroSieves™ allow the rapid gentle isolation of cells, organelles or cellular debris.
Specifications
Yes | |
For rapid collection of tissue culture cells without centrifugation | |
Matrix Filter CellMicroSieves | |
Nylon |