missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Concanavalin A, FITC-Conjugated
£92.00
Specifications
Concentration | 5μg/mL |
---|---|
Formulation | 0.05M Tris, 0.15M NaCl, 0.004M CaCl2, pH 7 to 7.2, contains Sodium Azide as a preservative |
Name | Rabbit Concanavalin A, FITC Conjugated from Jack bean |
pH Range | 7 to 7.2 |
Purification Method | Gel Filtration |
Specifications
5μg/mL | |
Rabbit Concanavalin A, FITC Conjugated from Jack bean | |
Gel Filtration | |
0°C | |
Rabbit |
0.05M Tris, 0.15M NaCl, 0.004M CaCl2, pH 7 to 7.2, contains Sodium Azide as a preservative | |
7 to 7.2 | |
Jack Bean | |
FITC | |
Lyophilized |