Learn More
MP Biomedicals™ Concanavalin A, FITC-Conjugated
Brand: MP Biomedicals™ 0215324505
Code : NEW
Additional Details : Weight : 0.00010kg
Specifications
5μg/mL | |
Rabbit Concanavalin A, FITC Conjugated from Jack bean | |
Gel Filtration | |
Jack Bean | |
FITC | |
Lyophilized |
0.05M Tris, 0.15M NaCl, 0.004M CaCl2, pH 7 to 7.2, contains Sodium Azide as a preservative | |
7 to 7.2 | |
5mg | |
0°C | |
Rabbit |
Safety and Handling
- Concanavalin A, FITC Conugated
Signal Word
- Danger
Hazard Category
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Concanavalin A