missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human MERTK (cMER), GST Tag Recombinant Protein
Human MERTK (cMER), GST Tag Recombinant Protein
£434.00 - £6900.00
Specifications
Accession Number | NP_006334 |
---|---|
Gene Alias | MERTK (cMER), MER |
KinaseFamily | AXL Kinase Family |
Species | Human |
Recombinant | Recombinant |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15458473
|
Invitrogen™
PR6317A |
100μg |
£1604.00
100µg |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10321833
|
Invitrogen™
PV4112 |
1mg |
£6900.00
1mg |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11583477
|
Invitrogen™
PV3627 |
10 μg |
£434.00
10µg |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
MERTK (cMER) is involved in the retinal pigment epithelium (RPE) phagocytosis pathway, which is implicated in human retinal disease.
Specifications
NP_006334 | |
AXL Kinase Family | |
Recombinant | |
Baculovirus |
MERTK (cMER), MER | |
Human | |
GST Tag |