missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human MERTK (cMER), GST Tag Recombinant Protein
Human MERTK (cMER), GST Tag Recombinant Protein
Brand: Invitrogen™ PV4112
Code : NEW
Additional Details : Weight : 0.00900kg
Description
MERTK (cMER) is involved in the retinal pigment epithelium (RPE) phagocytosis pathway, which is implicated in human retinal disease.
Specifications
NP_006334 | |
50mM Tris-HCl pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
1mg | |
RUO - research use only | |
MERTK (cMER), MER | |
Human | |
GST Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
63.4kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Cytoplasmic | |
AXL Kinase Family | |
Recombinant | |
Baculovirus |