missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LRRK2 (WT) Recombinant Protein
Mutations in the leucine-rich repeat kinase 2 (LRRK2) gene, which codes for a multi-domain protein kinase, are considered the most common genetic cause of late-onset autosomal-dominant Parkinson's disease.
£504.00 - £11100.00
Specifications
Accession Number | NP_940980.3 |
---|---|
For Use With (Application) | Kinase Assay |
Gene ID (Entrez) | 120892 |
Molecular Weight (g/mol) | 288 kDa |
Gene Alias | AURA17, DARDARIN, PARK8, RIPK7, ROCO2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15393806
|
Invitrogen™
A15198 |
100μg |
£2410.00
100µg |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15383806
|
Invitrogen™
A15197 |
10μg |
£504.00
10µg |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15303816
|
Invitrogen™
A15199 |
1mg |
£11100.00
1mg |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This full-length, purified and active wild-type LRRK2 protein may be useful in research to better understand the native protein, unaffected by mutations, as well as to explore the mechanisms underlying Parkinson's disease.
- Increase disease relevance by interrogating a purified and active full-length LRRK2 protein
- Uncover novel compounds or hits specific to the full-length form
- Advance Parkinson's research using the full-length protein and validated assays
Specifications
NP_940980.3 | |
120892 | |
AURA17, DARDARIN, PARK8, RIPK7, ROCO2 | |
Human | |
Store at -68 to -85°C. Aliquot at first use to avoid multiple freeze-thaws. If properly stored, this product is guaranteed for 6 months from date of purchase. | |
Kinases & Inhibitors | |
Liquid |
Kinase Assay | |
288 kDa | |
LRRK Kinase Family | |
DYKDDDDK | |
Baculovirus | |
Recombinant, Full Length | |
Neurobiology |