Learn More
Invitrogen™ LRRK2 (WT) Recombinant Protein
Mutations in the leucine-rich repeat kinase 2 (LRRK2) gene, which codes for a multi-domain protein kinase, are considered the most common genetic cause of late-onset autosomal-dominant Parkinson's disease.
Brand: Invitrogen™ A15199
Code : NEW
Additional Details : Weight : 0.00900kg
Description
This full-length, purified and active wild-type LRRK2 protein may be useful in research to better understand the native protein, unaffected by mutations, as well as to explore the mechanisms underlying Parkinson's disease.
- Increase disease relevance by interrogating a purified and active full-length LRRK2 protein
- Uncover novel compounds or hits specific to the full-length form
- Advance Parkinson's research using the full-length protein and validated assays
Specifications
NP_940980.3 | |
120892 | |
1mg | |
LRRK Kinase Family | |
DYKDDDDK | |
Baculovirus | |
Recombinant, Full Length | |
Neurobiology |
Kinase Assay | |
288 kDa | |
AURA17, DARDARIN, PARK8, RIPK7, ROCO2 | |
Human | |
Store at -68 to -85°C. Aliquot at first use to avoid multiple freeze-thaws. If properly stored, this product is guaranteed for 6 months from date of purchase. | |
Kinases & Inhibitors | |
Liquid |