missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LRRK2 [I2020T] Human Protein
LRRK2 is a multidomain protein that includes a kinase domain, a GTPase domain, and a leucine-rich repeat (LRR) domain.
£2555.00 - £12060.00
Specifications
Accession Number | NP_940980.2 |
---|---|
For Use With (Application) | Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology |
Molecular Weight (g/mol) | 204.9kDa |
Storage Requirements | Kinase proteins are shipped on dry ice and should be stored at -80°C. |
Regulatory Status | RUO - research use only |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15458683
|
Invitrogen™
PR8755A |
100μg |
£2555.00
100µg |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15478793
|
Invitrogen™
PV5855 |
1mg |
£12060.00
1mg |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Mutations in the LRRK2 gene have been associated with Parkinson 's disease, including the I2020T mutation in the kinase catalytic domain.
Specifications
NP_940980.2 | |
204.9kDa | |
RUO - research use only | |
LRRK2 I2020T | |
Human | |
Baculovirus |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
Kinase proteins are shipped on dry ice and should be stored at -80°C. | |
Catalytic | |
LRRK Kinase Family | |
GST Tag | |
Frozen |