Learn More
Invitrogen™ LRRK2 [I2020T] Human Protein
LRRK2 is a multidomain protein that includes a kinase domain, a GTPase domain, and a leucine-rich repeat (LRR) domain.
Brand:  Invitrogen™ PV5855
Code : NEW
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15478793
  • £12060.00 / 1mg
Estimated Shipment: 06-06-2024
to see stock.
Description
Description
Mutations in the LRRK2 gene have been associated with Parkinson 's disease, including the I2020T mutation in the kinase catalytic domain.
Specifications
Specifications
NP_940980.2
204.9kDa
Kinase proteins are shipped on dry ice and should be stored at -80°C.
Catalytic
LRRK Kinase Family
GST Tag
Frozen
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology
1mg
RUO - research use only
LRRK2 I2020T
Human
Baculovirus