missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hoefer™ TE Blotting Unit Replacement Parts and Accessories: Buffer Chamber
For use with TE62 Standard Transfer Tank with Cooling Chamber
£645.67 - £1354.80
Product Code | Brand | For Use With (Equipment) | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Includes | Price | Quantity & Availability | ||||
11584444
|
Hoefer™
TE67 |
TE 62 or the discontinued TE 62X Transfer Tank | Fittings, crochet hook and holder |
£1354.80
Each |
Estimated Shipment: 01-11-2024
Log in to see stock. |
||||
11564444
|
Hoefer™
TE56 |
TE 42 or discontinued TE 52X Transfer Tank | Crochet hook and holder |
£645.67
Each |
Estimated Shipment: 01-11-2024
Log in to see stock. |
||||
Description
- With Heat Exchanger
Specifications
Buffer Chamber |