missing translation for 'onlineSavingsMsg'
Learn More

Hoefer™ TE Blotting Unit Replacement Parts and Accessories: Buffer Chamber

For use with TE62 Standard Transfer Tank with Cooling Chamber

Brand:  Hoefer™ TE56

 View more versions of this product

Product Code. 11564444

  • £645.67 / Each
Estimated Shipment: 01-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • With Heat Exchanger
Specifications

Specifications

TE 42 or discontinued TE 52X Transfer Tank
Buffer Chamber
Crochet hook and holder
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers