missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ HDPE Standardized Screw Caps

Constructed of high-density polyethylene, one of the most versitile plasitcs, making it lightweight yet very strong. Kartell™ HDPE Standardized Screw Caps are used to tightly seal a Kartell™ PE Bottle, reducing the risk of spillage.

£16.65 - £23.70

Specifications

Color White
Material HDPE
Screw Material HDPE
Product Line Cap
Product Type HDPE Screw Cap
View More Specs

Products 2
Product Code Brand Closure Size Price Quantity & Availability  
Product Code Brand Closure Size Price Quantity & Availability  
12697765
View Documents Promotion Details
Kartell™
1580000
GL 18
£23.70
Pack of 100
Estimated Shipment: 11-02-2025
to see stock.
Add to basket
 
10673472
View Documents Promotion Details
Kartell™
1584000
GL 25
£16.65
Pack of 50
Estimated Shipment: 11-02-2025
to see stock.
Add to basket
 
Description

Description

  • Made of HDPE (High-density Polyethylene)
  • For use with PE bottles DIN 168
  • Easy to use screw cap
Specifications

Specifications

White
HDPE
HDPE Screw Cap
Screw Cap
HDPE
Cap
HDPE
Videos
SDS
Documents

Documents

Product Certifications