missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ HDPE Standardized Screw Caps

Product Code. 12697765
Click to view available options
Closure Size:
GL 18
GL 25
This item is not returnable. View return policy
This item is not returnable. View return policy

Constructed of high-density polyethylene, one of the most versitile plasitcs, making it lightweight yet very strong. Kartell™ HDPE Standardized Screw Caps are used to tightly seal a Kartell™ PE Bottle, reducing the risk of spillage.

  • Made of HDPE (High-density Polyethylene)
  • For use with PE bottles DIN 168
  • Easy to use screw cap

Specifications

Color White
Material HDPE
Screw Material HDPE
Product Line Cap
Product Type HDPE Screw Cap
For Use With (Equipment) Kartell™ Bottles 15801 - 15821
Closure Material HDPE
Closure Size GL 18
Closure Type Screw Cap
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'