missing translation for 'onlineSavingsMsg'
Learn More

Merck Millipore Super-Q™ Plus Water Purification System Accessories

For Super-Q Systems

£1280.00

Specifications

Capacity (Metric) 56 cm
Length (English) 22 in.
Length (Metric) 56 cm
View More Specs

Products 1
Product Code Brand Type Price Quantity & Availability  
Product Code Brand Type Price Quantity & Availability  
10468122
View Documents Promotion Details
Merck Millipore
CDFC02203
Ion Exchange Cartridge
£1280.00
Pack of 3
View alternative products
Save Up to 
»
Estimated Shipment: 11-07-2024
to see stock.
Add to basket
 
Description

Description

For Super-Q systems
Specifications

Specifications

56 cm
22 in.
56 cm
Videos
SDS
Documents

Documents

Product Certifications