missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck Millipore Super-Q™ Plus Water Purification System Accessories
For Super-Q Systems
Brand: Merck Millipore CDFC02203
Code : NEW
Additional Details : Weight : 0.50000kg
Description
For Super-Q™ systemsSpecifications
Water Purification System | |
22 in. | |
22 in. |
Ion Exchange Cartridge | |
56 cm | |
56 cm |