missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck Express™ PLUS Membrane Filters
Plain white polyethersulfone membranes provide ultrafast filtration for a variety of applications
Brand: Merck HPWP14250
Code : NEW
Additional Details : Weight : 0.10000kg
Description
Plain white polyethersulfone membranes provide ultrafast filtration for a variety of applications
The Millipore Express™ PLUS membrane provides ultrafast filtration of tissue culture media, additives, buffers and other aqueous solutions. This high throughput, low protein-binding membrane is also used in many of our ready-to-use sterile filtration devices.Features & Benefit:•Ultra fast filtration and high throughput shorten process time •Can be sterilized by autoclave, ethylene oxide, or gamma irradiation for sterile filtration applications Applications:Sterile Filtration, Buffer Filtration, Tissue Culture Media FiltrationSpecifications
Not Autoclavable | |
Screen Filter | |
0.45 μm | |
23 (at 690 Mbar (10 psi)) | |
Hydrophilic | |
Hydrophilic | |
142 mm |
White | |
PES | |
Non-sterile | |
Provide ultrafast filtration of tissue culture media, additives, buffers and other aqueous solutions. This membrane is also used in many ready-to-use sterile filtration devices. | |
Filter Membranes | |
142 mm |