missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck Express™ PLUS Membrane Filters
Plain white polyethersulfone membranes provide ultrafast filtration for a variety of applications
£122.00 - £672.00
Specifications
Autoclavable | Not Autoclavable |
---|---|
Color | White |
Filter Type | Screen Filter |
Material | PES |
Sterility | Non-sterile |
Product Code | Brand | Diameter (Metric) Filter | Pore Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Filter | Pore Size | Price | Quantity & Availability | ||||
10228250
|
Merck
GPWP04700 |
47 mm | 0.22 μm |
£363.00
Pack of 100 |
Estimated Shipment: 16-05-2024
Log in to see stock. |
||||
10400041
|
Merck
GPWP01300 |
13 mm | 0.22 μm |
£135.00
Pack of 100 |
Estimated Shipment: 14-06-2024
Log in to see stock. |
||||
10569022
|
Merck
GPWP02500 |
25 mm | 0.22 μm |
£208.00
Pack of 100 |
Estimated Shipment: 14-05-2024
Log in to see stock. |
||||
10054590
|
Merck
GPWP09050 |
90 mm | 0.22 μm |
£271.00
Pack of 50 |
Estimated Shipment: 18-06-2024
Log in to see stock. |
||||
10722151
|
Merck
GPWP14250 |
142 mm | 0.22 μm |
£672.00
Pack of 50 |
Estimated Shipment: 13-05-2024
Log in to see stock. |
||||
10022433
|
Merck
HPWP01300 |
13 mm | 0.45 μm |
£122.00
Pack of 100 |
Estimated Shipment: 14-06-2024
Log in to see stock. |
||||
10409183
|
Merck
HPWP14250 |
142 mm | 0.45 μm |
£535.00
Pack of 50 |
Estimated Shipment: 17-06-2024
Log in to see stock. |
||||
10764974
|
Merck
HPWP09050 |
90 mm | 0.45 μm |
£302.00
Pack of 50 |
Estimated Shipment: 13-05-2024
Log in to see stock. |
||||
10408023
|
Merck
HPWP02500 |
25 mm | 0.45 μm |
£167.00
Pack of 100 |
Estimated Shipment: 17-06-2024
Log in to see stock. |
||||
10326353
|
Merck
HPWP04700 |
47 mm | 0.45 μm |
£333.00
Pack of 100 |
Estimated Shipment: 13-05-2024
Log in to see stock. |
||||
Descrizione
Plain white polyethersulfone membranes provide ultrafast filtration for a variety of applications
- Hydrophilic, high-throughput, low-protein-binding
- Thickness: ≥150μm and ≤200μm
- Sterilization: by autoclave (121°C at 1 bar), EO or gamma
- Bacterial endotoxins: 0.5 EU/mL
- Gravimetric extractables: <1.5%
- Flowrate: 40mL/min. cm2 at 2 bar, 25°C
Applications:
Provide ultrafast filtration of tissue culture media, additives, buffers and other aqueous solutions. This membrane is also used in many ready-to-use sterile filtration devices.
Specifica
Not Autoclavable | |
Screen Filter | |
Non-sterile | |
Hydrophilic | |
Hydrophilic |
White | |
PES | |
Provide ultrafast filtration of tissue culture media, additives, buffers and other aqueous solutions. This membrane is also used in many ready-to-use sterile filtration devices. | |
Filter Membranes |