missing translation for 'onlineSavingsMsg'
Learn More

Merck Express™ PLUS Membrane Filters

Plain white polyethersulfone membranes provide ultrafast filtration for a variety of applications

Brand:  Merck HPWP02500

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10408023

  • £167.00 / Pack of 100
Estimated Shipment: 28-06-2024
to see stock.
Add to basket

Description

Description

Plain white polyethersulfone membranes provide ultrafast filtration for a variety of applications

The Millipore Express™ PLUS membrane provides ultrafast filtration of tissue culture media, additives, buffers and other aqueous solutions. This high throughput, low protein-binding membrane is also used in many of our ready-to-use sterile filtration devices.Features & Benefit:•Ultra fast filtration and high throughput shorten process time •Can be sterilized by autoclave, ethylene oxide, or gamma irradiation for sterile filtration applications Applications:Sterile Filtration, Buffer Filtration, Tissue Culture Media Filtration
Specifications

Specifications

Not Autoclavable
Screen Filter
0.45 μm
23 (at 690 Mbar (10 psi))
Hydrophilic
Hydrophilic
25 mm
White
PES
Non-sterile
Provide ultrafast filtration of tissue culture media, additives, buffers and other aqueous solutions. This membrane is also used in many ready-to-use sterile filtration devices.
Filter Membranes
25 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers